Palladium(II) sulphide 99% (≥72% Pd)
Leverantör: Thermo Scientific
Varning
Synonyms: Palladium sulphide, Palladium monosulphide
12674.03 12674.06
12674.03EA 5850 SEK
12674.03 12674.06
Palladium(II) sulphide 99% (≥72% Pd)
Palladium(II) sulphide
Purity: 99% CAS No.: 12125-22-3 Molecular Formula: PdS Formula Weight: 138.46 Physical Form: Crystalline Density: 6.60 Melting Point: 950° dec. MDL No.: MFCD00016285
Formel: PdS
MW: 138,49 g/mol
Smältpunkt: 950 °C (dec.)
Densitet: 6,60
Storage Temperature: Omgivande
MDL Number: MFCD00016285
CAS nummer: 12125-22-3
EINECS: 235-190-6