Cesium hydroxide 50% (w/w) i vattenlösning , 99.9%
Leverantör: Thermo Fisher Scientific
Fara
10020.ND 10020.NF
10020.NDEA 3875 SEK
10020.ND 10020.NF
Cesium hydroxide 50% (w/w) i vattenlösning , 99.9%
Cesium hydroxide
Purity: 99.9% CAS No.: 21351-79-1 Molecular Formula: CsOH Formula Weight: 149.91 Physical Form: Liquid Density: 1.725 Appearance: Colourless Odour: Odourless UN No.: 2681 MDL No.: MFCD00010964
Formel: CsOH
MW: 149.91 g/mol
Smältpunkt: 338.85
Densitet: 1.725
Storage Temperature: Omgivande
MDL Number: MFCD00010964
CAS nummer: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8 II