Borane dimethyl sulphide complex 2 M i THF
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Borane-methyl sulphide complex
Packaged under argon in resealable ChemSeal™ bottles.
42965.AE 42965.AP
42965.AEEA 2075 SEK
42965.AE 42965.AP
Borane dimethyl sulphide complex 2 M i THF
Borane dimethyl sulphide complex
CAS No.: 13292-87-0 Molecular Formula: C2H9BS Formula Weight: 75.97 Flash Point: -17°(-1°F) Physical Form: Liquid Density: 0.855 Appearance: Colourless to pale yellow Odour: Characteristic Beilstein Reference: 3663489 UN No.: 3399 MDL No.: MFCD00013189
Formel: C₂H₉BS
MW: 75.97 g/mol
Smältpunkt: -27
Densitet: 0.855
Flampunkt: –17 °C (1 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013189
CAS nummer: 13292-87-0
EINECS: 236-313-6
UN: 3399
ADR: 4.3 II