Borane dimethyl sulphide complex 2 M I toluen
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Borane-methyl sulphide complex
Packaged under argon in resealable ChemSeal™ bottles.
42963.AE 42963.AP
42963.AEEA 3250 SEK
42963.AE 42963.AP
Borane dimethyl sulphide complex 2 M I toluen
Borane dimethyl sulphide complex
CAS No.: 13292-87-0 Molecular Formula: C2H9BS Formula Weight: 75.97 Flash Point: 7°(45°F) Physical Form: Liquid Density: 0.856 Appearance: Colourless Beilstein Reference: 3663489 UN No.: 3399 MDL No.: MFCD00013189
Formel: C₂H₉BS
MW: 75.97 g/mol
Smältpunkt: -27
Densitet: 0.856
Flampunkt: 7 °C (45 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013189
CAS nummer: 13292-87-0
EINECS: 236-313-6
UN: 3399
ADR: 4.3 I