Refractive Index Liquids, Series A
Leverantör: CARGILLE
1809Y1.458 1809Y1.500 1809Y1.550 1809Y1.600 1809Y1.640
CARG1809Y1.458EA 5775 SEK
CARG1809Y1.458 CARG1809Y1.500 CARG1809Y1.550 CARG1809Y1.600 CARG1809Y1.640
Refractive Index Liquids, Series A
Standarder Standarder för brytningsindex
Refractive index liquids have a broad range of applications in diversified fields.
  • Colourless to light yellow liquid
  • Slight odour
Adjustment: ±0,0002
Choose from range nD 1,460 to 1,640