4-Cholesten-3-one ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: (8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3(2H)-one, (+)-4-Cholesten-3-one
A11743.06 A11743.14
A11743.06EA 1650 SEK
A11743.06 A11743.14
4-Cholesten-3-one ≥98%
4-Cholesten-3-one
Purity: 98+% CAS No.: 601-57-0 Molecular Formula: C27H44O Formula Weight: 384.65 Melting Point: 79-81° Beilstein Reference: 4707774
Formel: C₂₇H₄₄O
MW: 384.65 g/mol
Smältpunkt: 79…81 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003663
CAS nummer: 601-57-0
EINECS: 210-005-1