2-Aminoisobutyric acid 98+%
Leverantör: Thermo Scientific
Synonyms: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid
225760250 225761000
ACRO225760250EA 665 SEK
ACRO225760250 ACRO225761000
2-Aminoisobutyric acid 98+%
2-Aminoisobutyric acid
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Kokpunkt: 204 °C (1013 hPa)
Smältpunkt: 335 °C
Densitet: 1,278 g/cm³ (20 °C)
MDL Number: MFCD00008049
CAS nummer: 62-57-7
EINECS: 200-544-0
Merck Index: 13,00444
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.0 %