4,4 '-Bi-o-toluidin dihydroklorid ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: o-Toluidine dihydrochloride
A18196.14 A18196.22
A18196.14EA 1275 SEK
A18196.14 A18196.22
4,4 '-Bi-o-toluidin dihydroklorid ≥97%
4,4 '-Bi-o-toluidin dihydroklorid
Purity: 97% CAS No.: 612-82-8 Molecular Formula: C14H16N2.2HCl Formula Weight: 285.22 Odour: Odourless Melting Point: 300° UN No.: 3077 MDL No.: MFCD00012960
Formel: C₁₄H₁₈Cl₂N₂
MW: 285,22 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012960
CAS nummer: 612-82-8
EINECS: 210-322-5
ADR: 9,III