2-Amino-3-bromo-5-nitrobenzonitrile

Leverantör: Thermo Scientific

SB01878EE
MAYBSB01878EEEA 0 SEK
MAYBSB01878EE
2-Amino-3-bromo-5-nitrobenzonitrile
2-Amino-3-bromo-5-nitrobenzonitrile
Formel: C₇H₄BrN₃O₂
MW: 242,03 g/mol
Smältpunkt: 233…235 °C
Densitet: 1,949 g/cm³ (–123 °C)
MDL Number: MFCD00054185
CAS nummer: 17601-94-4
EINECS: 241-574-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR