L-alanin, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
1.01700.1000 1.01700.9010
1.01700.1000EA 17900 SEK
1.01700.1000 1.01700.9010
L-alanin, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
L-alanin
Formel: C₃H₇NO₂
MW: 89.09 g/mol
Kokpunkt: 189
Smältpunkt: 79
Densitet: 1.4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064410
CAS nummer: 56-41-7
EINECS: 200-273-8
Merck Index: 13,00203