L-Fenylalanin ≥99%
Leverantör: Thermo Fisher Scientific
Synonyms: L-α-Fenylalanin
A13238.0B A13238.14 A13238.18 A13238.22 A13238.36
A13238.0BEA 13300 SEK
A13238.0B A13238.14 A13238.18 A13238.22 A13238.36
L-Fenylalanin ≥99%
L-Fenylalanin
Purity: 99% CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Cryst powder Appearance: White Melting Point: ca 275° dec. Beilstein Reference: 1910408 Merck Reference: 14,7271 Optical Rotation: -34° (c=2 in water) MDL No.: MFCD00064227
Formel: C₉H₁₁NO₂
Kokpunkt: 313 °C (1013 hPa)
Smältpunkt: ∼ 275 °C (dec.)
Densitet: 0,754 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00064227
CAS nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,7271