Aluminiumoxid aktiverad neutral Brockmann Grade I, Supelco®
Leverantör: Merck
M-Clarity™ quality level = MQ200
06300-25KG 06300-5KG
06300-25KGEA 18390 SEK
06300-25KG 06300-5KG
Aluminiumoxid aktiverad neutral Brockmann Grade I, Supelco®
Aluminium oxide activated neutral
Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration. Aluminum oxides are generally used as solid support phases in solid phase extraction (SPE)-based clean-up procedures employed in chromatographic methods
Formel: Al₂O₃
MW: 101.96 g/mol
Smältpunkt: 2040 °C (lit.)
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6