Fumaronitrile ≥98%
Leverantör: Thermo Fisher Scientific
Fara
L14911.03 L14911.06
L14911.03EA 620 SEK
L14911.03 L14911.06
Fumaronitrile ≥98%
Fumaronitrile
Purity: 98+% CAS No.: 764-42-1 Molecular Formula: C4H2N2 Formula Weight: 78.07 Odour: Recognizable Melting Point: 94-99° Boiling Point: 186° Beilstein Reference: 969245 UN No.: 3439 MDL No.: MFCD00001928
Formel: C₄H₂N₂
MW: 78.07 g/mol
Kokpunkt: 186 °C
Smältpunkt: 94…99 °C
Densitet: 0.94
Storage Temperature: Omgivande
MDL Number: MFCD00001928
CAS nummer: 764-42-1
EINECS: 212-122-3
UN: 3439
ADR: 6.1 III