Svavelsyra 500 ml/l (∼65,1%)
Leverantör: FF-CHEMICALS
|
Fara
|
FF21691000
FF21695000
FFCHFF21691000EA
1675
SEK
FFCHFF21691000
FFCHFF21695000
Svavelsyra 500 ml/l (∼65,1%)
Svavelsyra
Formel:
H₂SO₄ MW: 98,08 g/mol Kokpunkt: ∼100 °C (1013 hPa) Densitet: 1,5…1.8 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00064589 CAS nummer: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...