Saltsyra 6 mol/l (6 N)

Leverantör: FF CHEMICALS
Varning

Synonyms: Hydrochloric acid solution, Muriatic acid

FF2320500 FF2321000
FFCHFF2320500EA 480 SEK
FFCHFF2320500 FFCHFF2321000
Saltsyra 6 mol/l (6 N)
Saltsyra
Formel: HCl
MW: 36,46 g/mol
Densitet: 1,06 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR