Polypropylenglykol, M.W. 1,000
Leverantör: Thermo Fisher Scientific
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
40812.18 40812.30
40812.18EA 870 SEK
40812.18 40812.30
Polypropylenglykol, M.W. 1,000
Polypropylenglykol
CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Physical Form: Liquid Appearance: Colourless MDL No.: MFCD01779683
Formel: H(OCH₂CH₂CH₂)nOH
MW: 1025
Storage Temperature: Omgivande
MDL Number: MFCD00084446
CAS nummer: 25322-69-4
EINECS: 500-039-8