Sodium hexanitritocobaltate(III) ACS, Fluka™
Leverantör: Honeywell Chemicals
Varning
Synonyms: Trinatriumhexanitritokobaltat
233722-100G 233722-500G
233722-100GEA 3350 SEK
233722-100G 233722-500G
Sodium hexanitritocobaltate(III) ACS, Fluka™
Sodium hexanitritocobaltate(III)
Formel: Na₃Co(NO₂)₆
MW: 403,94 g/mol
Smältpunkt: 220 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003512
CAS nummer: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1,II
Merck Index: 13,08677
Order Now
Specifikation testresultat
Appearance of the substance complying
Insoluble in acetic acid Max. 0.020 %
Suitability for K determination complying