Sodium chlorodifluoroacetate 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic acid sodium salt
404590250 404590050 404591000
ACRO404590250EA 1150 SEK
ACRO404590250 ACRO404590050 ACRO404591000
Sodium chlorodifluoroacetate 96%
Sodium chlorodifluoroacetate
Formel: C₂ClF₂NaO₂
MW: 152.46 g/mol
Smältpunkt: 195…201 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064771
CAS nummer: 1895-39-2
EINECS: 217-586-0
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥95 %