Sodium chlorodifluoroacetate 97%, stavar
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium 2-chloro-2,2-difluoroacetate, Chlorodifluoroacetic acid sodium salt
A15756.14 A15756.22 A15756.36
A15756.14EA 1525 SEK
A15756.14 A15756.22 A15756.36
Sodium chlorodifluoroacetate 97%, stavar
Sodium chlorodifluoroacetate
Purity: 97% CAS No.: 1895-39-2 Molecular Formula: ClCF2CO2Na Formula Weight: 152.46 Melting Point: ca 195° dec. Beilstein Reference: 4162363
Formel: C₂ClF₂NaO₂
MW: 152.46 g/mol
Smältpunkt: ∼ 195 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00064771
CAS nummer: 1895-39-2
EINECS: 217-586-0