ε-Hexalactone monomer 99%
Leverantör: Thermo Scientific
Varning
Synonyms: ε-Caprolactone, Oxepan-2-one, Hexan-6-olide
173440010 173442500 173440025
ACRO173440010EA 2425 SEK
ACRO173440010 ACRO173442500 ACRO173440025
ε-Hexalactone monomer 99%
ε-Hexalactone
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 225 °C (1013 hPa)
Smältpunkt: –1 °C
Densitet: 1,03 g/cm³
Flampunkt: 109 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003267
CAS nummer: 502-44-3
EINECS: 207-938-1
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4640 (20°C, 589 nm)
Water ≤0.1 %
GC ≥98.5 %