(±)-γ-Undecalactone 98%
Leverantör: Thermo Fisher Scientific
Varning
259490250 259491000 259495000
ACRO259490250EA 440 SEK
ACRO259490250 ACRO259491000 ACRO259495000
(±)-γ-Undecalactone 98%
(±)-γ-Undecalactone
Formel: C₁₁H₂₀O₂
Kokpunkt: 286 °C (760 mmHg)
Densitet: 0,9425 g/cm³
Flampunkt: 145 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005405
CAS nummer: 104-67-6
EINECS: 203-225-4
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.939 to 0.948