4-Acetylphenylboronic acid ≥98%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
(4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane
B23234.03
B23234.06
B23234.14
B23234.03EA
805
SEK
B23234.03
B23234.06
B23234.14
4-Acetylphenylboronic acid ≥98%
4-Acetylphenylboronic acid
Purity: 98%
CAS No.: 149104-90-5
Molecular Formula: C8H9BO3
Formula Weight: 163.97
Melting Point: ca 240° dec.
Beilstein Reference: 7869162
Formel:
C₈H₉BO₃ MW: 163,97 g/mol Smältpunkt: ∼ 240 °C (dec.) Storage Temperature: Omgivande |
MDL Number:
MFCD01074667 CAS nummer: 149104-90-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...