5-Bromonicotinonitrile
Leverantör: Thermo Scientific
Synonyms:
5-Bromopyridine-3-carbonitrile, 3-Bromopyridine-5-carbonitrile, 5-Bromo-3-cyanopyridine, 3-Bromo-5-cyanopyridine
KM08398DA
KM08398EA
MAYBKM08398DAEA
0
SEK
MAYBKM08398DA
MAYBKM08398EA
5-Bromonicotinonitrile
5-Bromonicotinonitrile
Formel:
C₆H₃BrN₂ MW: 183,01 g/mol |
MDL Number:
MFCD00174363 CAS nummer: 35590-37-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...