3-Aminophenylboronic acid monohydrate 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate
352760010 352760050
ACRO352760010EA 910 SEK
ACRO352760010 ACRO352760050
3-Aminophenylboronic acid monohydrate 98%
3-Aminophenylboronic acid monohydrate
Formel: C₆H₈BNO₂·1H₂O
MW: 154.96 g/mol
Smältpunkt: 94…97 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00149554
CAS nummer: 206658-89-1
EINECS: 250-189-0
Order Now
Specifikation testresultat
Appearance Beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %