(±)-Citronellal, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: (+/-)-Citronellal, (±)-3,7-Dimethyloct-6-enal, (+/-)-3,7-Dimethyl-6-octenal
M-Clarity™ quality level = MQ200
8.14575.0100 8.14575.0500
8.14575.0100EA 608 SEK
8.14575.0100 8.14575.0500
(±)-Citronellal, Sigma-Aldrich®
(±)-Citronellal
Formel: C₁₀H₁₈O
Kokpunkt: 207 °C (1013 hPa)
Smältpunkt: –15 °C
Densitet: 0,8548 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00038090
CAS nummer: 106-23-0
EINECS: 203-376-6
UN: 3082
ADR: 9,III
Merck Index: 13,02353
Order Now
Specifikation testresultat
Solubility (20 °C) practically insoluble
Molar Mass 154.25 g/mol
Boiling Point 208 °C (1013 hPa)
Vapor Pressure 14 hPa (88 °C)
Flash Point 77 °C
Refractive Index 1.446 (20 °C)
Explosion Limit 1.2 - 4.5 %(V)
Density 0.85 g/cm3 (20 °C)
pH 7 (H2O)
Ignition Point 202 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material