2-Methoxynicotinic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid
A13193.03 A13193.06
A13193.03EA 830 SEK
A13193.03 A13193.06
2-Methoxynicotinic acid ≥98%
2-Methoxynicotinic acid
Purity: 98% CAS No.: 16498-81-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 147-151° MDL No.: MFCD00085110
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Smältpunkt: 147…151 °C
Storage Temperature: Omgivande
MDL Number: MFCD00085110
CAS nummer: 16498-81-0