4,4'-Isopropylidendifenol ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Bisfenol A
A10324.0B A10324.0I A10324.30
A10324.0BEA 1175 SEK
A10324.0B A10324.0I A10324.30
4,4'-Isopropylidendifenol ≥97%
4,4'-Isopropylidendifenol
Purity: 97+% CAS No.: 80-05-7 Molecular Formula: C15H16O2 Formula Weight: 228.29 Flash Point: 227°(440°F) Melting Point: 153-158° Boiling Point: 200°/4mm Beilstein Reference: 1107700 Merck Reference: 14,1297 MDL No.: MFCD00002366
Formel: C₁₅H₁₆O₂
MW: 228.29 g/mol
Kokpunkt: 200 °C (4 mmHg)
Smältpunkt: 153…158 °C
Densitet: 1.205 g/cm³ (20 °C)
Flampunkt: 227 °C (440 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002366
CAS nummer: 80-05-7
EINECS: 201-245-8
Merck Index: 13,01296