1,4-Phenyldiboronic acid 96%
Leverantör: Thermo Scientific
Varning
Synonyms: p-Phenylenebisboronic acid, 1,4-Phenylenediboronic acid, 1,4-Phenyldiboric acid, 1,4-Benzenediboronic acid
B24064.03 B24064.06 B24064.14
B24064.03EA 990 SEK
B24064.03 B24064.06 B24064.14
1,4-Phenyldiboronic acid 96%
1,4-Phenyldiboronic acid
Purity: 96% CAS No.: 4612-26-4 Molecular Formula: C6H8B2O4 Formula Weight: 165.75 Melting Point: 300° Beilstein Reference: 2836921
Formel: C₆H₈B₂O₄
MW: 165,75 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00236018
CAS nummer: 4612-26-4