Adiponitrile ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Hexanedinitrile
B23122.AK B23122.AU
B23122.AKEA 800 SEK
B23122.AK B23122.AU
Adiponitrile ≥98%
Adiponitrile
Purity: 98% CAS No.: 111-69-3 Molecular Formula: NC(CH2)4CN Formula Weight: 108.14 Flash Point: 163°(325°F) Refractive Index: 1.4380 Odour: Odourless Melting Point: 2° Boiling Point: 295° Beilstein Reference: 1740005 UN No.: 2205 MDL No.: MFCD00001975
Formel: C₆H₈N₂
MW: 108.14 g/mol
Kokpunkt: 295 °C
Smältpunkt: 2
Densitet: 0.951
Flampunkt: 163 °C (325 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001975
CAS nummer: 111-69-3
EINECS: 203-896-3
UN: 2205
ADR: 6.1 III