2,6-Bis(hydroxymethyl)-p-cresol ≥90%, tech.
Leverantör: Thermo Fisher Scientific
Varning
A17369.14 A17369.22 A17369.36
A17369.14EA 1025 SEK
A17369.14 A17369.22 A17369.36
2,6-Bis(hydroxymethyl)-p-cresol ≥90%, tech.
2,6-Bis(hydroxymethyl)-p-cresol
Purity: 90% CAS No.: 91-04-3 Molecular Formula: C9H12O3 Formula Weight: 168.19 Melting Point: 119-126° Beilstein Reference: 1240237
Formel: C₉H₁₂O₃
Kokpunkt: 363 °C (1013 hPa)
Smältpunkt: 119…126 °C
Densitet: 1,382 g/cm³ (–173 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004619
CAS nummer: 91-04-3
EINECS: 202-036-4