2-(2-Metoxietoxi)etanol ≥98% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether
Stabilization: Stabilised med BHT/Ionol
A16063.0F A16063.AP
A16063.0FEA 1500 SEK
A16063.0F A16063.AP
2-(2-Metoxietoxi)etanol ≥98% stabilised
2-(2-Metoxietoxi)etanol
Stabilized with 50-150mg/kg BHT
Purity: 98% CAS No.: 111-77-3 Molecular Formula: C5H12O3 Formula Weight: 120.15 Flash Point: 83°(181°F) Refractive Index: 1.4260 Odour: Light Melting Point: -70° Boiling Point: 194° Beilstein Reference: 1697812 Merck Reference: 14,6037 MDL No.: MFCD00002871
Formel: C₅H₁₂O₃
MW: 120.15 g/mol
Kokpunkt: 194 °C
Smältpunkt: –70 °C
Densitet: 1.010
Flampunkt: 83 °C (181 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002871
CAS nummer: 111-77-3
EINECS: 203-906-6
Merck Index: 14,6037