JALAS® GRAN PREMIO 1518 ANTISLIP+ ESD Safety Sandals
Leverantör: EJENDALS
1518-49 1518-48 1518-50
EJEN1518-49EA 2005 SEK
EJEN1518-49 EJEN1518-48 EJEN1518-50
JALAS® GRAN PREMIO 1518 ANTISLIP+ ESD Safety Sandals
Skor och kängor
Sandal that breathes, with a spacious last. With all the necessary safety features for those who prioritize comfort and cool feet. The sandal is ventilated and has a spacious last. Hook and loop closure, nail protection in steel and toe cap in aluminium.
  • ProNose toe reinforcement
  • Wide fit
  • Heat-resistant outsole
  • Oil-resistant outsole
  • Anti-static properties
  • Ventilating insole
  • Double shock absorption zones
  • Conforms with IEC 61340-5-1 (ESD)
These sandals has highest level of protection, excellent grip, perfect fit, extra comfortable, excellent shock absorption and very breathable.
Outsole material: Nitrile rubber outsole
Upper material: PU-coated leather
Insole: FX2 Pro-X
Pronose material: PU
Toecap material: Aluminium
Lining material: Polyester, polyamide
Nail Protection material: Steel
Colour: Black, grey, yellow
Net weight: 1,23 kg
Certifikat: S1, P, SRC, HRO.