Tetrapropylammonium perruthenate
Leverantör: TCI
Varning
T1559-1G T1559-5G
TCIAT1559-1GEA 1575 SEK
TCIAT1559-1G TCIAT1559-5G
Tetrapropylammonium perruthenate
Tetrapropylammonium perruthenate
Formel: C₁₂H₂₈NO₄Ru
Smältpunkt: 163°C
Storage Temperature: Kylskåp
MDL Number: MFCD00074914
CAS nummer: 114615-82-6
UN: 1479
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e44b3f3670647a0cda7ab28f5b9d18fbc39ebb59&vid=e44b3f3670647a0cda7ab28f5b9d18fbc39ebb59&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Dark green to Dark blue to Black powder to crystal
Functionality test to pass test