3-Methylfuran ≥98.0% stabilised
Leverantör: TCI EUROPE
Fara
Stabilization: med hydrokinon
M0939-1G M0939-5G
TCIAM0939-1GEA 2325 SEK
TCIAM0939-1G TCIAM0939-5G
3-Methylfuran ≥98.0% stabilised
3-Methylfuran
Formel: C₅H₆O
MW: 82,1 g/mol
Kokpunkt: 66°C
Densitet: 0,923 g/cm³ (18 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00060134
CAS nummer: 930-27-8
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8e2d0326c7ea9249bb8366d7182ff29b562ea40&vid=a8e2d0326c7ea9249bb8366d7182ff29b562ea40&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %