4-Chloropyrocatechol ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 4-Chloro-1,2-dihydroxybenzene, 4-Chlorobenzene-1,2-diol, 4-Chlorocatechol

C0568-25G C0568-5G
TCIAC0568-25GEA 4825 SEK
TCIAC0568-25G TCIAC0568-5G
4-Chloropyrocatechol ≥98.0%
4-Chloropyrocatechol
Formel: C₆H₅ClO₂
MW: 144,56 g/mol
Kokpunkt: 140°C
Smältpunkt: 90…91 °C
MDL Number: MFCD00059614
CAS nummer: 2138-22-9
EINECS: 218-381-9
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0467e5eb45ca0defae4fb208f5003d8ee21906b&vid=e0467e5eb45ca0defae4fb208f5003d8ee21906b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 88.0 to 93.0 deg-C
Solubility in Methanol ithin almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR