N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Supelco®
Leverantör: Merck
Fara
Synonyms: BSTFA+TMCS (99:1)
M-Clarity™ quality level = MQ100
T6381-100G T6381-10AMP T6381-1AMP T6381-25G T6381-5G
T6381-100GEA 10220 SEK
T6381-100G T6381-10AMP T6381-1AMP T6381-25G T6381-5G
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, Supelco®
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane
Formel: C₈H₁₈F₃NOSi₂
MW: 257.40 g/mol
MDL Number: MFCD00008269
CAS nummer: 25561-30-2
EINECS: 247-103-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2179fa263117895b1e29bb713687b18d721aec1&vid=4e4790fd424bcfac7bdb26c06aab912c21307f3b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=486b948309c5919be5be9c458789a5ddfe5312a2&vid=744901561fbfc992b9b8826b7d31b88cfe1eb9bb&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96ba19fdf727f35b85ab7754a4cbbf79815e583a&vid=e8b07c3bf4c2bf0daeaac78acf2d6d97c4d2e3ee&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85f4632feea09fb8105b818bd652f5871f73a7a9&vid=d0f4d8b1b6a608b248ca0e9eee050641ee8f73d0&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74b1601cf2907295967998bd7e33f876414e1595&vid=6f494e5a6b292514f885bb71487122779766a432&prid=web&clid=SAPDAM