N-metyl-N-(trimetylsilyl) trifluoracetamid, Supelco®
Leverantör: Merck
Fara
Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide
M7891-25G M7891-5G
M7891-25GEA 2630 SEK
M7891-25G M7891-5G
N-metyl-N-(trimetylsilyl) trifluoracetamid, Supelco®
N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Formel: C₆H₁₂F₃NOSi
MW: 199,25 g/mol
Kokpunkt: 130 °C (1013 hPa)
Densitet: 1,07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS nummer: 24589-78-4
EINECS: 246-331-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a0715dae01d88ca2eb3929384f2f0c16831c688&vid=9a0715dae01d88ca2eb3929384f2f0c16831c688&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88a9d1582b58668d76c99be4fc9b4e17da5be019&vid=88a9d1582b58668d76c99be4fc9b4e17da5be019&prid=web&clid=SAPDAM