N-metyl-N-(trimetylsilyl) trifluoracetamid, Supelco®
Leverantör: Merck
Fara
Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide
M-Clarity™ quality level = MQ100
69479-25ML 69479-5ML 69479-10X1ML
69479-25MLEA 8840 SEK
69479-25ML 69479-5ML 69479-10X1ML
N-metyl-N-(trimetylsilyl) trifluoracetamid, Supelco®
N-metyl-N-(trimetylsilyl) trifluoracetamid
Formel: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Kokpunkt: 130
Densitet: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00000411
CAS nummer: 24589-78-4
EINECS: 246-331-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d146999835387e9eb6175715ba5453f576eac&vid=4c5df68c2bb042a7be46f8d847bba9b247818475&prid=web&clid=SAPDAM