Chromium hexacarbonyl, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: Chromium carbonyl, Hexacarbonylchromium, Hexacarbonylchromium(0), Chromium(0) hexacarbonyl
M-Clarity™ quality level = MQ200
8.22196.0010 8.22196.0025
8.22196.0010EA 3460 SEK
8.22196.0010 8.22196.0025
Chromium hexacarbonyl, Sigma-Aldrich®
Chromium hexacarbonyl
Formel: C₆CrO₆
Kokpunkt: 210 °C (1013 hPa)
Smältpunkt: 90 °C
Densitet: 1,77 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010945
CAS nummer: 13007-92-6
EINECS: 235-852-4
UN: 3466
ADR: 6.1,III
Merck Index: 13,02253
Order Now
Specifikation testresultat
Solubility (20 °C) insoluble
Melting Point 149 - 150 °C
Molar Mass 220.06 g/mol
Boiling Point 151 °C (1013 hPa)
Vapor Pressure 0.35 hPa (20 °C)
Density 1.77 g/cm3 (20 °C)