(2,4-Diklorfenoxi)ättiksyra, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: 2,4-D
M-Clarity™ quality level = MQ200
8.20451.0005 8.20451.0250 8.20451.1000 8.20451.9020
8.20451.0005EA 317 SEK
8.20451.0005 8.20451.0250 8.20451.1000 8.20451.9020
(2,4-Diklorfenoxi)ättiksyra, Sigma-Aldrich®
(2,4-Diklorfenoxi)ättiksyra
Formel: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
Kokpunkt: 346
Smältpunkt: 137…139 °C
Densitet: 1.563 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004300
CAS nummer: 94-75-7
EINECS: 202-361-1
Merck Index: 13,02825
Order Now
Specifikation testresultat
Solubility 0.3 g/l (25 °C)
Melting Point 134 - 137 °C
Molar Mass 221.04 g/mol
Bulk Density 720 kg/m3
Boiling Point 160 °C (0.53 hPa)
Density 1.42 g/cm3 (25 °C)
Synonyms 2,4 D-acid
Hill Formula C8H6Cl2O3
Chemical Formula 2,4-(Cl)2C6H3OCH2COOH