N, O-bis (trimetylsilyl)acetamid, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
M-Clarity™ quality level = MQ200
8.18727.0025 8.18727.0100
8.18727.0025EA 644 SEK
8.18727.0025 8.18727.0100
N, O-bis (trimetylsilyl)acetamid, Sigma-Aldrich®
N, O-bis (trimetylsilyl)acetamid
Formel: C₈H₂₁NOSi₂
Kokpunkt: 162 °C (1013 hPa)
Smältpunkt: ∼24 °C
Densitet: 0,82 g/cm³ (20 °C)
Flampunkt: 40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008270
CAS nummer: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8 (3),II
Order Now
Specifikation testresultat
Solubility (20 °C) decomposes
Molar Mass 203.43 g/mol
Boiling Point 43 °C (11 hPa)
Vapor Pressure 10 hPa (50 °C)
Flash Point 40 °C
Density 0.83 g/cm3 (20 °C)