Natriumklorid, EMPROVE® api BP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Saline, NaCl, Sea salt, Halite
1.06444.1000 1.06444.9029
1.06444.1000EA 1425 SEK
1.06444.1000 1.06444.9029
Natriumklorid, EMPROVE® api BP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient, SAFC®
Natriumklorid
Formel: NaCl
Kokpunkt: 1413 °C (1013 hPa)
Smältpunkt: 801 °C
Densitet: 2,16 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003477
CAS nummer: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599