Oleyl alcohol ≥99%, clear, colourless liquid
Leverantör: MP Biomedicals
Varning
Synonyms: cis-Octadec-9-en-1-ol, cis-9-Octadecen-1-ol
0215599694 0215599690 0215599691
ICNA0215599694EA 8600 SEK
ICNA0215599694 ICNA0215599690 ICNA0215599691
Oleyl alcohol ≥99%, clear, colourless liquid
Oleyl alcohol
Oleyl alcohol is a long-chain aliphatic alcohol that occurs naturally in fish oils. It is prepared by synthetic reduction of plant-derived oleic acid.
Oleyl alcohol is used in largescale applications as the softening and lubrication of textile fabrics, and the production of carbon paper, stencil paper, and printing ink. Oleyl alcohol is also utilized as an antifoam agent and cutting lubricant. It also is a precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. Oleyl alcohol has also been incorporated into various formulations for drug delivery.
Formel: C₁₈H₃₆O
MW: 268,48 g/mol
Kokpunkt: 207 °C (lit.)
Smältpunkt: 13…19 °C (lit.)
Densitet: 0,85 g/cm³ (20 °C)
Storage Temperature: Frys
MDL Number: MFCD00002993
CAS nummer: 143-28-2
EINECS: 205-597-3
Merck Index: 12,06968