2-(4-jodofenyl)-3-(4-nitrofenyl)-5-fenyltetrazoliumklorid ≥99%, yellow powder
Leverantör: MP Biomedicals
Varning
Synonyms: Iodonitrotetrazolium chloride, p-Iodonitrotetrazolium violet, INT, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazol-3-ium chloride, Iodonitrotetrazolium violet, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyltetrazolium chloride
0210206625 0210206605 02102066.1 02102066.5 0210206601
ICNA0210206625EA 25400 SEK
ICNA0210206625 ICNA0210206605 ICNA02102066.1 ICNA02102066.5 ICNA0210206601
2-(4-jodofenyl)-3-(4-nitrofenyl)-5-fenyltetrazoliumklorid ≥99%, yellow powder
2-(4-jodofenyl)-3-(4-nitrofenyl)-5-fenyltetrazoliumklorid
p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases.
p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases.
Soluble in 5% methanol/water(1:1)(clear yellow); slightly soluble in water, alcohol, DMSO.
Formel: C₁₉H₁₃ClIN₅O₂
MW: 505.7 g/mol
Smältpunkt: 227…229 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00011961
CAS nummer: 146-68-9
EINECS: 205-676-2