1-Chloro-5-fluoro-2-iodobenzene

Leverantör: Thermo Scientific

Synonyms: 2-Chloro-4-fluoro-1-iodobenzene, 2-Chloro-4-fluoroiodobenzene

TL00223EE
MAYBTL00223EEEA 0 SEK
MAYBTL00223EE
1-Chloro-5-fluoro-2-iodobenzene
1-Chloro-5-fluoro-2-iodobenzene
Formel: C₆H₃ClFI
MW: 256,45 g/mol
MDL Number: MFCD00051774
CAS nummer: 101335-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR