1-Amino-2-methylbutane

Leverantör: Thermo Scientific

Synonyms: 2-Methylbutylamine, 2-Methyl-1-butylamine

SB01376DA SB01376EA SB01376.50
MAYBSB01376DAEA 0 SEK
MAYBSB01376DA MAYBSB01376EA MAYBSB01376.50
1-Amino-2-methylbutane
1-Amino-2-methylbutane
Formel: C₅H₁₃N
MW: 87.16 g/mol
Kokpunkt: 97
Densitet: 0.755 g/cm³ (18 °C)
MDL Number: MFCD00008147
CAS nummer: 96-15-1
EINECS: 202-483-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR