EDTA disodium salt, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverantör: Avantor
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
7124.1000EA 640 SEK
7124.1000 7124.9020
EDTA disodium salt, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt
Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336,21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00070672
CAS nummer: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556
Order Now
Specifikation testresultat
Titer (mol/l) 0.0997 - 0.1003