4-Bromo-2,6-dichlorobenzenesulphonyl chloride 95%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Bromo-2,6-dichlorobenzene-1-sulphonyl chloride
L19879.03 L19879.06
L19879.03EA 1200 SEK
L19879.03 L19879.06
4-Bromo-2,6-dichlorobenzenesulphonyl chloride 95%
4-Bromo-2,6-dichlorobenzenesulphonyl chloride
Purity: 95% CAS No.: 351003-54-8 Molecular Formula: C6H2BrCl3O2S Formula Weight: 324.41 Flash Point: 110°(230°F) Melting Point: 42-48° UN No.: 3261 MDL No.: MFCD03094651
Formel: C₆H₂BrCl₃O₂S
MW: 324,41 g/mol
Smältpunkt: 42…48 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD03094651
CAS nummer: 351003-54-8
UN: 3261
ADR: 8,II