2,6-Bis(trifluoromethyl)pyridine ≥97%
Leverantör: Thermo Fisher Scientific
Varning
L19446.03 L19446.MD
L19446.03EA 1800 SEK
L19446.03 L19446.MD
2,6-Bis(trifluoromethyl)pyridine ≥97%
2,6-Bis(trifluoromethyl)pyridine
Purity: 97% CAS No.: 455-00-5 Molecular Formula: C7H3F6N Formula Weight: 215.10 Flash Point: 67°(153°F) Melting Point: 55-58° Boiling Point: 82°/18mm Beilstein Reference: 188806 MDL No.: MFCD00236675
Formel: C₇H₃F₆N
MW: 215.1 g/mol
Kokpunkt: 82 °C (18 mmHg)
Smältpunkt: 55…58 °C
Flampunkt: 67 °C (153 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00236675
CAS nummer: 455-00-5