3-Methoxybenzenesulphonyl chloride ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 3-Methoxybenzene-1-sulphonyl chloride
L17050.03 L17050.06
L17050.03EA 2000 SEK
L17050.03 L17050.06
3-Methoxybenzenesulphonyl chloride ≥97%
3-Methoxybenzenesulphonyl chloride
Purity: 97% CAS No.: 10130-74-2 Molecular Formula: C7H7ClO3S Formula Weight: 206.65 Flash Point: 110°(230°F) Refractive Index: 1.5570 Boiling Point: 158-159°/20mm Beilstein Reference: 2093871 UN No.: 3265 MDL No.: MFCD01318155
Formel: C₇H₇ClO₃S
Kokpunkt: 158…159 °C (20 mmHg)
Densitet: 1,460
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD01318155
CAS nummer: 10130-74-2
UN: 3265
ADR: 8,II